(±)-2-Phenyl-3-butyn-2-ol ≥98%

Synonyms: 2-Phenylbut-3-yn-2-ol

212997-5G 212997-100G 212997-25G
220043-968EA 43.08 USD
220043-968 220043-966 220043-967
(±)-2-Phenyl-3-butyn-2-ol ≥98%
(±)-2-Phenyl-3-butyn-2-ol
Formula: C₁₀H₁₀O
MW: 146.19 g/mol
MDL Number: MFCD00004454
CAS Number: 127-66-2
EINECS: 204-855-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR