Europium(III) chloride hexahydrate ≥99.9%

Synonyms: Europium(III) chloride hydrate, Europium trichloride hexahydrate

212881-5G 212881-25G
220043-950EA 167.69 USD
220043-950 220043-949
Europium(III) chloride hexahydrate ≥99.9%
Europium(III) chloride hexahydrate
Formula: Cl₃Eu·6H₂O
MW: 366.41 g/mol
Melting Pt: 158 °C
Density: 4.89 g/cm³ (20 °C)
MDL Number: MFCD00149701
CAS Number: 13759-92-7
EINECS: 233-040-4

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR