2-Amino-5-nitrobenzophenone ≥98%

Synonyms: (2-Amino-5-nitrophenyl)(phenyl)methanone

211737-25G 211737-100G
220042-224EA 57.08 USD
220042-224 220042-223
2-Amino-5-nitrobenzophenone ≥98%
2-Amino-5-nitrobenzophenone
Formula: C₁₃H₁₀N₂O₃
MW: 242.23 g/mol
Melting Pt: 163.5 °C
Density: 1.474 g/cm³ (20 °C)
MDL Number: MFCD00007364
CAS Number: 1775-95-7
EINECS: 217-207-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR