(±)-Nipecotic acid

Synonyms: H-DL-Nip-OH, (±)-3-Piperidinecarboxylic acid, Piperidine-3-carboxylic acid, Nipecotic acid

211672-5G 211672-1G 211672-25G
220043-142EA 34.77 USD
220043-142 220043-140 220043-141
(±)-Nipecotic acid
(±)-Nipecotic acid
Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD00005992
CAS Number: 498-95-3
EINECS: 207-873-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR