Lanthanum(III) chloride hydrate ≥99.9%

Synonyms: Lanthanum trichloride hydrate

211605-500G 211605-100G
220043-776EA 690.77 USD
220043-776 220043-775
Lanthanum(III) chloride hydrate ≥99.9%
Lanthanum(III) chloride hydrate
Formula: Cl₃La·H₂O
MW: 263.28 g/mol
Boiling Pt: 1000 °C (1013 hPa)
Melting Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00149753
CAS Number: 20211-76-1
EINECS: 233-237-5

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR