Gadolinium trinitrate hexahydrate ≥99.9%

Synonyms: Gadolinium (III) nitrate hexahydrate

211591-100G 211591-25G
220043-772EA 395.38 USD
220043-772 220043-773
Gadolinium trinitrate hexahydrate ≥99.9%
Gadolinium trinitrate hexahydrate
Formula: GdN₃O₉·6H₂O
MW: 451.36 g/mol
Melting Pt: 91.5 - 92 °C
Density: 2.4 g/cm³ (20 °C)
MDL Number: MFCD00149728
CAS Number: 19598-90-4
EINECS: 233-437-2
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR