Methyl-p-benzoquinone

Warning

Synonyms: 2-Methylcyclohexa-2,5-diene-1,4-dione, Methylquinone, Methyl-1,4-benzoquinone

211311-5G 211311-100G
220041-176EA 73.09 USD
220041-176 220041-175
Methyl-p-benzoquinone
Methyl-p-benzoquinone
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: 65…67 °C
Density: 1.08 g/cm³ (75.5 °C)
MDL Number: MFCD00001603
CAS Number: 553-97-9
EINECS: 209-056-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR