9-Phenanthrenol, Technical Grade

Warning

Synonyms: 9-Hydroxyphenanthrene, Phenanthren-9-ol, 9-Phenanthrol

211281-1G 211281-5G
220041-171EA 202.46 USD
220041-171 220041-172
9-Phenanthrenol, Technical Grade
9-Phenanthrenol
Formula: C₁₄H₁₀O
MW: 194.23 g/mol
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 155 °C
MDL Number: MFCD00082298
CAS Number: 484-17-3
EINECS: 207-602-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR