4-Phenoxybenzaldehyde ≥98%

Synonyms: p-Phenoxybenzenecarbaldehyde, 4-Phenoxybenzenecarbaldehyde

211265-25G 211265-5G
220043-660EA 331.69 USD
220043-660 220041-591
4-Phenoxybenzaldehyde ≥98%
4-Phenoxybenzaldehyde
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 24.5 °C
Density: 1.1522 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003383
CAS Number: 67-36-7
EINECS: 200-650-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR