Boron trichloride 1 M in hexane

211249-100ML 211249-800ML
220043-658EA 76.38 USD
220043-658 220043-659
Boron trichloride 1 M in hexane
Boron trichloride
Formula: BCl₃
MW: 117.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011313
CAS Number: 10294-34-5
EINECS: 233-658-4
UN: 3286
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR