Lead(II) bromide ≥98%

Danger

Synonyms: Lead dibromide, Lead(II) bromide

211141-100G 211141-500G
220043-642EA 160 USD
220043-642 220043-643
Lead(II) bromide ≥98%
Lead(II) bromide
Formula: Br₂Pb
MW: 367.01 g/mol
Boiling Pt: 916 °C (1013 hPa)
Melting Pt: 362 °C
Density: 6.676 g/cm³ (0 °C)
MDL Number: MFCD00011156
CAS Number: 10031-22-8
EINECS: 233-084-4
UN: 2291
ADR: 6.1,III
Merck Index: 12,05418

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR