4-tert-Butylaniline ≥99%

Synonyms: 4-(tert-Butyl)aniline

209864-5G 209864-25G
220043-113EA 67.42 USD
220043-113 220043-112
4-tert-Butylaniline ≥99%
4-tert-Butylaniline
Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 15 °C
Density: 0.941 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007899
CAS Number: 769-92-6
EINECS: 212-215-9
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR