Selenium ≥99.5%

Danger

209651-250G 209651-50G
220043-569EA 263.08 USD
220043-569 220043-570
Selenium ≥99.5%
Selenium
Formula: Se
Boiling Pt: 684.9 °C (1013 hPa)
Melting Pt: 217 °C
Density: 4.81 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134090
CAS Number: 7782-49-2
EINECS: 231-957-4
UN: 3283
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR