(3-Methoxyphenyl)acetonitrile ≥99%

Synonyms: 3-Methoxybenzyl cyanide, 2-(3-Methoxyphenyl)acetonitrile

209392-25G 209392-5G
220043-540EA 221.85 USD
220043-540 220043-541
(3-Methoxyphenyl)acetonitrile ≥99%
(3-Methoxyphenyl)acetonitrile
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 164 °C (1013 hPa)
Density: 1.05 g/cm³ (20 °C)
MDL Number: MFCD00001910
CAS Number: 19924-43-7
EINECS: 243-428-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR