3-Methoxybenzyl chloride ≥97%

Danger

Synonyms: 1-(Chloromethyl)-3-methoxybenzene, 3-(Chloromethyl)anisole

209384-5G 209384-25G
220043-539EA 24.77 USD
220043-539 220043-538
3-Methoxybenzyl chloride ≥97%
3-Methoxybenzyl chloride
Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 124 °C (1013 hPa)
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000907
CAS Number: 824-98-6
EINECS: 212-541-1
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR