Titanium(II) hydride

Warning

Synonyms: Titanium hydride, Titanium dihydride

209279-500G 209279-100G
220043-530EA 401.23 USD
220043-530 220043-529
Titanium(II) hydride
Titanium(II) hydride
Formula: H₂Ti
MW: 49.9 g/mol
Density: 3.91 g/cm³ (20 °C)
MDL Number: MFCD00151301
CAS Number: 7704-98-5
EINECS: 231-726-8
UN: 1871
ADR: 4.1,II
Merck Index: 14,09473

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR