Copper(II) sulfate hydrate

Warning

Synonyms: Cupric sulphate hydrate

209201-25G 209201-1KG
220043-055EA 34.34 USD
220043-055 220043-054
Copper(II) sulfate hydrate
Copper(II) sulphate hydrate
Formula: CuO₄S·H₂O
MW: 177.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149682
CAS Number: 23254-43-5
EINECS: 231-847-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR