Copper(II) sulfate pentahydrate ACS

Warning

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

209198-250G 209198-100G 209198-500G 209198-2.5KG 209198-5G
220043-045EA 46.15 USD
220043-045 220043-043 220043-046 220043-044 220043-047
Copper(II) sulfate pentahydrate ACS
Copper(II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR