Potassium osmate(VI) dihydrate

Synonyms: Potassium osmium(VI) oxide dihydrate, Potassium dioxidodioxoosmium dihydrate, Dipotassium osmate dihydrate

209104-100MG 209104-500MG 209104-2.5G
220043-510EA 82.77 USD
220043-510 220043-512 220043-511
Potassium osmate(VI) dihydrate
Potassium osmate(VI) dihydrate
Formula: K₂O₄Os·2H₂O
MW: 368.45 g/mol
MDL Number: MFCD00149919
CAS Number: 10022-66-9
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR