Germanium(IV) chloride ≥99.99%

Synonyms: Germanium tetrachloride, Tetrachlorogermane

208450-5G 208450-25G
220043-460EA 193.85 USD
220043-460 220043-459
Germanium(IV) chloride ≥99.99%
Germanium(IV) chloride
Formula: Cl₄Ge
MW: 214.42 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: -52 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011029
CAS Number: 10038-98-9
EINECS: 233-116-7
UN: 3264
ADR: 8,II
Merck Index: 12,04418

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR