Magnesium chloride

208337-100G 208337-1KG 208337-5KG
220042-413EA 47.23 USD
220042-413 220042-414 220042-415
Magnesium chloride
Magnesium chloride
Formula: MgCl₂
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 715 °C
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR