Vanadium(III) chloride

Warning

208272-25G 208272-1G 208272-5G
220042-676EA 250.77 USD
220042-676 220042-675 220042-677
Vanadium(III) chloride
Vanadium(III) chloride
Formula: Cl₃V
MW: 157.3 g/mol
Density: 3 g/cm³ (20 °C)
MDL Number: MFCD00011454
CAS Number: 7718-98-1
EINECS: 231-744-6
UN: 2475
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR