Zinc chloride ≥98%, Reagent Grade, Sigma-Aldrich®
|
Danger
|
Synonyms:
Zinc dichloride, Dichlorozinc, Zinc (II) chloride
M-Clarity™ quality level = MQ100
208086-5G
208086-100G
208086-500G
220043-422EA
15.26
USD
220043-422
220043-420
220043-421
Zinc chloride ≥98%, Reagent Grade, Sigma-Aldrich®
Zinc chloride
Formula:
ZnCl₂ MW: 136.3 g/mol Boiling Pt: 732 °C (1013 hPa) Melting Pt: 283 °C Density: 2.91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8,III Merck Index: 13,10185 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...