Europium trinitrate pentahydrate ≥99.9%

Synonyms: Europium (III) nitrate pentahydrate

207918-10G 207918-1G 207918-50G
220043-333EA 513.85 USD
220043-333 220043-334 220043-335
Europium trinitrate pentahydrate ≥99.9%
Europium trinitrate pentahydrate
Formula: EuN₃O₉·5H₂O
MW: 428.05 g/mol
CAS Number: 63026-01-7
EINECS: 233-380-3
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR