Potassium thiocyanate ≥99.0% ACS

Warning

207799-100G 207799-2.5KG
220043-311EA 72 USD
220043-311 220043-309 220043-310
Potassium thiocyanate ≥99.0% ACS
Potassium thiocyanate
Formula: KSCN
Boiling Pt: 500 °C (1013 hPa)
Melting Pt: 175 °C
Density: 1.61 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR