4-Ethynylanisole ≥97%

Synonyms: (4-Methoxyphenyl)ethyne, (4-Methoxyphenyl)acetylene, 1-Ethynyl-4-methoxybenzene, 1-Methoxy-4-ethynylbenzene

206490-1G 206490-5G
220043-257EA 117.38 USD
220043-257 220043-258
4-Ethynylanisole ≥97%
4-Ethynylanisole
Formula: C₉H₈O
MW: 132.16 g/mol
MDL Number: MFCD00168815
CAS Number: 768-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR