Diammonium tetrachloroplatinate

Danger

Synonyms: Ammonium tetrachloroplatinate(II), Ammonium tetrachloroplatinate

206105-5G 206105-1G
220043-149EA 960 USD
220043-149 220043-148
Diammonium tetrachloroplatinate
Diammonium tetrachloroplatinate
Formula: H₈Cl₄N₂Pt
MW: 372.97 g/mol
Melting Pt: 140 °C
Density: 2.93 g/cm³ (20 °C)
MDL Number: MFCD00010885
CAS Number: 13820-41-2
EINECS: 237-499-1
UN: 3288
ADR: 6.1,III
Merck Index: 12,00583

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR