Hexachloroplatinic(IV) acid hexahydrate ACS

Danger

Synonyms: Hydrogen hexachloroplatinate (IV) hexahydrate, Chloroplatinic acid hexahydrate, Dihydrogen hexachloroplatinate (IV) hexahydrate

206083-25G 206083-1G
220042-501EA 1526.15 USD
220042-501 220042-500 220042-502
Hexachloroplatinic(IV) acid hexahydrate ACS
Hexachloroplatinic(IV) acid hexahydrate
Formula: H₂PtCl₆·6H₂O
MW: 517.9 g/mol
Melting Pt: 150 °C
Density: 2.434 g/cm³ (20 °C)
MDL Number: MFCD00149910
CAS Number: 18497-13-7
EINECS: 241-010-7
UN: 2507
ADR: 8,III
Merck Index: 13,07609

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR