Platinium(IV) oxide

Warning

Synonyms: Platinum dioxide, Adam's type catalyst

206032-5G 206032-1G 206032-250MG
220041-924EA 446.54 USD
220041-924 220042-388 220042-389
Platinium(IV) oxide
Platinium(IV) oxide
Formula: O₂Pt
MW: 227.08 g/mol
Melting Pt: 450 °C
Density: 11.8 g/cm³ (20 °C)
MDL Number: MFCD00011184
CAS Number: 1314-15-4
EINECS: 215-223-0
UN: 1479
ADR: 5.1,II
Merck Index: 12,07686

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR