Palladium (II) sulfate ≥98%

205788-5G 205788-1G
220043-094EA 856.92 USD
220043-094 220043-093
Palladium (II) sulfate ≥98%
Palladium(II) sulphate
Formula: O₄PdS
MW: 202.48 g/mol
MDL Number: MFCD00011173
CAS Number: 13566-03-5
EINECS: 236-957-8
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR