Palladium(II) nitrate hydrate

205761-2G 205761-10G
220043-092EA 203.54 USD
220043-092 220043-091
Palladium(II) nitrate hydrate
Palladium(II) nitrate hydrate
Formula: N₂O₆Pd·xH₂O
MDL Number: MFCD00149818
CAS Number: 207596-32-5
EINECS: 233-265-8
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR