Bis(2-butoxyethyl)ether ≥99%

Synonyms: Diethylene glycol dibutyl ether, 2,2-Dibutoxyethyl ether

205621-100ML 205621-500ML 205621-1L
220043-066EA 67.09 USD
220043-066 220043-068 220043-067
Bis(2-butoxyethyl)ether ≥99%
Bis(2-butoxyethyl)ether
Formula: C₁₂H₂₆O₃
MW: 218.34 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: –60 °C
Density: 0.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009459
CAS Number: 112-73-2
EINECS: 204-001-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR