Dibutyltin dichloride

Synonyms: Dichlorodibutylstannane, Di-n-butyltin dichloride, DBTC

205494-250G 205494-50G
220042-988EA 74.08 USD
220042-988 220042-989
Dibutyltin dichloride
Dibutyltin dichloride
Formula: C₈H₁₈Cl₂Sn
MW: 303.85 g/mol
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 37 - 39 °C
Density: 1.37 to 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000518
CAS Number: 683-18-1
EINECS: 211-670-0
UN: 2928
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR