Tellurium tetrachloride ≥99%

Danger

205338-5G 205338-100G 205338-25G
220042-971EA 133.69 USD
220042-971 220042-969 220042-970
Tellurium tetrachloride ≥99%
Tellurium tetrachloride
Formula: Cl₄Te
MW: 269.41 g/mol
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 224 °C
Density: 3.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011262
CAS Number: 10026-07-0
EINECS: 233-055-6
UN: 3260
ADR: 8,II
Merck Index: 12,09279

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR