Lead(II) perchlorate hydrate ≥98%

205311-50G 205311-250G
220042-968EA 41.88 USD
220042-968 220042-967
Lead(II) perchlorate hydrate ≥98%
Lead(II) perchlorate hydrate
Formula: Cl₂O₈Pb·H₂O
MW: 424.12 g/mol
CAS Number: 207500-00-3
EINECS: 237-125-7
UN: 1470
ADR: 5.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR