Titanium(IV) isopropoxide

205273-2L 205273-500ML 205273-100ML
220042-958EA 175.15 USD
220042-958 220042-959 220042-957
Titanium(IV) isopropoxide
Titanium(IV) isopropoxide
Formula: C₁₂H₂₈O₄Ti
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 18…20 °C
Density: 0.9711 g/cm³ (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00008871
CAS Number: 546-68-9
EINECS: 208-909-6
UN: 2413
ADR: 3,III
Merck Index: 12,09614

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR