Praseodymium(III,IV) oxide ≥99.9%

Warning

205176-250G 205176-50G
220042-753EA 792.31 USD
220042-753 220042-754
Praseodymium(III,IV) oxide ≥99.9%
Praseodymium(III,IV) oxide
Formula: O₁₁Pr₆
MW: 1021.44 g/mol
Melting Pt: 2012…2072 °C
Storage Temperature: Ambient
MDL Number: MFCD00011178
CAS Number: 12037-29-5
EINECS: 234-857-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR