Diyttrium trioxide

Synonyms: Yttrium oxide, Yttrium (III) oxide

205168-250G 205168-10G 205168-50G
220042-751EA 613.85 USD
220042-751 220042-750 220042-752
Diyttrium trioxide
Diyttrium trioxide
Formula: Y₂O₃
MW: 225.81 g/mol
Boiling Pt: 4300 °C (1013 hPa)
Melting Pt: 2410 °C
Density: 5.01 g/cm³ (20 °C)
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR