Praseodymium trichloride hydrate ≥99.9%

Synonyms: Praseodymium (III) chloride hydrate

205141-10G 205141-50G
220042-748EA 124.77 USD
220042-748 220042-749
Praseodymium trichloride hydrate ≥99.9%
Praseodymium trichloride hydrate
Formula: Cl₃Pr·H₂O
MW: 265.28 g/mol
MDL Number: MFCD00149828
CAS Number: 19423-77-9
EINECS: 233-794-4

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR