Praseodymium(III) nitrate hexahydrate ≥99.9%

Synonyms: Praseodymium trinitrate hexahydrate

205133-50G 205133-250G 205133-10G
220042-480EA 238.46 USD
220042-480 220042-479 220042-478
Praseodymium(III) nitrate hexahydrate ≥99.9%
Praseodymium(III) nitrate hexahydrate
Formula: N₃O₉Pr·6H₂O
MW: 435.01 g/mol
Melting Pt: 56 °C
Density: 2.233 g/cm³ (20 °C)
MDL Number: MFCD00149826
CAS Number: 15878-77-0
EINECS: 233-796-5
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR