Tris(triphenylphosphine)rhodium(I) chloride

Synonyms: Wilkinson's catalyst, Chlorotris(triphenylphosphine)rhodium(I), Tris(triphenylphosphine)chlororhodium

205036-1G 205036-5G 205036-250MG
220042-472EA 241.23 USD
220042-472 220042-474 220042-473
Tris(triphenylphosphine)rhodium(I) chloride
Tris(triphenylphosphine)rhodium(I) chloride
Formula: C₅₄H₄₅ClP₃Rh
MW: 925.23 g/mol
Melting Pt: 157 °C
Density: 1.363 g/cm³ (20 °C)
MDL Number: MFCD00010016
CAS Number: 14694-95-2
EINECS: 238-744-5
Merck Index: 12,02226

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR