Zirconyl chloride hydrate ≥99.99% (trace metals basis)

Danger

Synonyms: Zirconium(IV) dichloride oxide hydrate, Zirconium dichloride oxide hydrate

205028-10G 205028-50G
220042-470EA 144.15 USD
220042-470 220042-471
Zirconyl chloride hydrate ≥99.99% (trace metals basis)
Zirconyl chloride hydrate
Formula: Cl₂OZr·H₂O
MW: 196.14 g/mol
MDL Number: MFCD00149897
CAS Number: 15461-27-5
EINECS: 231-717-9
UN: 3260
ADR: 8,II
Merck Index: 14,10184

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR