Zinc sulfate heptahydrate

Danger

204986-10G 204986-50G
220042-466EA 110.15 USD
220042-466 220042-467
Zinc sulfate heptahydrate
Zinc sulphate heptahydrate
Formula: ZnSO₄·7H₂O
MW: 287.54 g/mol
Melting Pt: 100 °C
Density: 1.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 12,10293

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR