Diyttrium trioxide

Synonyms: Yttrium oxide, Yttrium (III) oxide

204927-10G 204927-50G
220042-953EA 141.69 USD
220042-953 220042-954
Diyttrium trioxide
Diyttrium trioxide
Formula: Y₂O₃
MW: 225.81 g/mol
Boiling Pt: 4300 °C (1013 hPa)
Melting Pt: 2410 °C
Density: 5.01 g/cm³ (20 °C)
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR