Vanadyl sulfate hydrate

Warning

Synonyms: Vanadium (IV) sulphate oxide hydrate, Vanadium(IV) oxide sulphate hydrate, Vanadium oxide sulphate hydrate, Basic vanadium (IV) sulphate hydrate

204862-10G 204862-50G
220042-945EA 203.08 USD
220042-945 220042-946
Vanadyl sulfate hydrate
Vanadyl sulphate hydrate
Formula: O₅SV·H₂O
MW: 163.00 g/mol (anhy)
MDL Number: MFCD00149933
CAS Number: 123334-20-3
EINECS: 248-652-7
UN: 2931
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR