Tungsten trioxide

Warning

204781-50G 204781-10G
220042-938EA 724.62 USD
220042-938 220042-937
Tungsten trioxide
Tungsten trioxide
Formula: O₃W
MW: 231.84 g/mol
Melting Pt: 1473 °C
Density: 7.16 g/cm³ (20 °C)
MDL Number: MFCD00011466
CAS Number: 1314-35-8
EINECS: 215-231-4
Merck Index: 12,09947

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR