Titanium(IV) oxide

204757-25G 204757-125G
220042-936EA 172.31 USD
220042-936 220042-935
Titanium(IV) oxide
Titanium(IV) oxide
Formula: TiO₂
Boiling Pt: 2900 °C (1013 hPa)
Melting Pt: 1855 °C
Density: 4.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS Number: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR