Tin(II) chloride ≥99.99%

Danger

Synonyms: Stannous chloride, Tin dichloride

204722-10G 204722-50G
220042-928EA 120.62 USD
220042-928 220042-929
Tin(II) chloride ≥99.99%
Tin(II) chloride
Formula: SnCl₂
MW: 189.62 g/mol
Boiling Pt: 623 °C (1013 hPa)
Melting Pt: 246 °C
Density: 3.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011241
CAS Number: 7772-99-8
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 13,08861

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR