Terbium(III) chloride hexahydrate ≥99.999%

204560-1G 204560-5G
220042-914EA 153.69 USD
220042-914 220042-915
Terbium(III) chloride hexahydrate ≥99.999%
Terbium(III) chloride hexahydrate
Formula: Cl₃Tb·6H₂O
MW: 373.38 g/mol
Melting Pt: 145 °C
Density: 4.35 g/cm³ (20 °C)
MDL Number: MFCD00149870
CAS Number: 13798-24-8
EINECS: 233-132-4
Merck Index: 12,09300

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR