Tantalum(V) oxide

204536-10G 204536-50G
220042-908EA 170.77 USD
220042-908 220042-909
Tantalum(V) oxide
Tantalum(V) oxide
Formula: O₅Ta₂
Melting Pt: 1870…1880 °C
Density: 8.2 g/cm³ (20 °C)
MDL Number: MFCD00011254
CAS Number: 1314-61-0
EINECS: 215-238-2
Merck Index: 14,09056

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR